2-(2-Bromophenyl)-4'-cyanoacetophenone ≥97%

Supplier: Matrix Scientific

086716-1G 086716-2G 086716-5G
102850-198EA 592.31 USD
102850-198 102850-200 102850-202
2-(2-Bromophenyl)-4'-cyanoacetophenone ≥97%
2-(2-Bromophenyl)-4'-cyanoacetophenone

Hazard: Irritant

Formula: C₁₅H₁₀BrNO
MDL Number: MFCD02260722
CAS Number: 898784-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR