2-(2-Bromophenyl)-3'-cyanoacetophenone ≥97%
Supplier: Matrix Scientific
086715-1G
086715-2G
086715-5G
102850-192EA
592.31
USD
102850-192
102850-194
102850-196
2-(2-Bromophenyl)-3'-cyanoacetophenone ≥97%
2-(2-Bromophenyl)-3'-cyanoacetophenone
Hazard: Irritant
Formula:
C₁₅H₁₀BrNO |
MDL Number:
MFCD02260721 CAS Number: 898784-10-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...