2-(3-Cyanophenyl)acetophenone ≥97%

Supplier: Matrix Scientific

086713-1G 086713-2G 086713-5G
102850-180EA 886.15 USD
102850-180 102850-182 102850-184
2-(3-Cyanophenyl)acetophenone ≥97%
2-(3-Cyanophenyl)acetophenone

Hazard: Irritant

Formula: C₁₅H₁₁NO
Melting Pt: 74…77 °C
MDL Number: MFCD02260691
CAS Number: 62043-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR