2-(2-Cyanophenyl)acetophenone ≥97%

Supplier: Matrix Scientific

086712-1G 086712-2G 086712-5G
102850-174EA 592.31 USD
102850-174 102850-176 102850-178
2-(2-Cyanophenyl)acetophenone ≥97%
2-(2-Cyanophenyl)acetophenone

Hazard: Irritant

Formula: C₁₅H₁₁NO
MDL Number: MFCD00029561
CAS Number: 10517-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR