2-(4-Methoxyphenyl)acetophenone ≥97%
Supplier: Matrix Scientific
086711-1G
086711-2G
086711-5G
102850-168EA
538.46
USD
102850-168
102850-170
102850-172
2-(4-Methoxyphenyl)acetophenone ≥97%
2-(4-Methoxyphenyl)acetophenone
Hazard: Irritant
Formula:
C₁₅H₁₄O₂ MW: 226.27 g/mol |
MDL Number:
MFCD00096294 CAS Number: 24845-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...