2-(4-Bromophenyl)acetophenone ≥97%
Supplier: Matrix Scientific
086708-1G
086708-2G
086708-5G
102850-150EA
612.31
USD
102850-150
102850-152
102850-154
2-(4-Bromophenyl)acetophenone ≥97%
2-(4-Bromophenyl)acetophenone
Hazard: Irritant
Formula:
C₁₄H₁₁BrO Melting Pt: 147…149 °C |
MDL Number:
MFCD01851462 CAS Number: 22421-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...