ω-(4-Chlorophenyl)acetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(4-Chlorophenyl)acetophenone

086706-1G 086706-2G 086706-5G
102850-138EA 347.69 USD
102850-138 102850-140 102850-142
ω-(4-Chlorophenyl)acetophenone ≥97%
ω-(4-Chlorophenyl)acetophenone

Hazard: Irritant

Formula: C₁₄H₁₁ClO
MW: 230.6895 g/mol
MDL Number: MFCD02260686
CAS Number: 6332-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR