ω-(3-Chlorophenyl)acetophenone ≥97%

Supplier: Matrix Scientific

086705-1G 086705-2G 086705-5G
102850-132EA 612.31 USD
102850-132 102850-134 102850-136
ω-(3-Chlorophenyl)acetophenone ≥97%
ω-(3-Chlorophenyl)acetophenone

Hazard: Irritant

Formula: C₁₄H₁₁ClO
Melting Pt: 41-43°C
MDL Number: MFCD02260685
CAS Number: 27798-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR