ω-(4-Fluorophenyl)acetophenone ≥97%
Supplier: Matrix Scientific
086704-1G
086704-2G
086704-5G
102850-126EA
347.69
USD
102850-126
102850-128
102850-130
ω-(4-Fluorophenyl)acetophenone ≥97%
ω-(4-Fluorophenyl)acetophenone
Hazard: Irritant
Formula:
C₁₄H₁₁FO |
MDL Number:
MFCD02260684 CAS Number: 347-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...