ω-(3-Fluorophenyl)acetophenone ≥97%
Supplier: Matrix Scientific
086703-1G
086703-2G
086703-5G
102850-120EA
347.69
USD
102850-120
102850-122
102850-124
ω-(3-Fluorophenyl)acetophenone ≥97%
ω-(3-Fluorophenyl)acetophenone
Hazard: Irritant
Formula:
C₁₄H₁₁FO |
MDL Number:
MFCD02260683 CAS Number: 347-90-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...