2-(2-Fluorophenyl)acetophenone ≥95%

Supplier: Matrix Scientific

086702-1G 086702-2G 086702-5G
102850-114EA 347.69 USD
102850-114 102850-116 102850-118
2-(2-Fluorophenyl)acetophenone ≥95%
2-(2-Fluorophenyl)acetophenone

Hazard: Irritant

Formula: C₁₄H₁₁FO
MDL Number: MFCD02260682
CAS Number: 3826-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR