4'-Nitro-2-phenylacetophenone ≥97%
Supplier: Matrix Scientific
086701-1G
086701-2G
086701-5G
102850-108EA
581.54
USD
102850-108
102850-110
102850-112
4'-Nitro-2-phenylacetophenone ≥97%
4'-Nitro-2-phenylacetophenone
Hazard: Irritant
Formula:
C₁₄H₁₁NO₃ |
MDL Number:
MFCD02260681 CAS Number: 3769-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...