3'-Nitro-2-phenylacetophenone ≥97%

Supplier: Matrix Scientific

086700-1G 086700-2G 086700-5G
102850-102EA 575.38 USD
102850-102 102850-104 102850-106
3'-Nitro-2-phenylacetophenone ≥97%
3'-Nitro-2-phenylacetophenone

Hazard: Irritant

Formula: C₁₄H₁₁NO₃
MDL Number: MFCD02260680
CAS Number: 55251-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR