2-Phenyl-4'-(trifluormethyl)acetophenone ≥97%
Supplier: Matrix Scientific
086699-1G
086699-2G
086699-5G
102850-096EA
592.31
USD
102850-096
102850-098
102850-100
2-Phenyl-4'-(trifluormethyl)acetophenone ≥97%
2-Phenyl-4'-(trifluormethyl)acetophenone
Hazard: Irritant
Formula:
C₁₅H₁₁F₃O |
MDL Number:
MFCD02260678 CAS Number: 61062-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...