4'-Cyano-2-phenylacetophenone ≥97%

Supplier: Matrix Scientific

086697-1G 086697-2G 086697-5G
102850-084EA 749.23 USD
102850-084 102850-086 102850-088
4'-Cyano-2-phenylacetophenone ≥97%
4'-Cyano-2-phenylacetophenone

Hazard: Irritant

Formula: C₁₅H₁₁NO
MDL Number: MFCD02260675
CAS Number: 60694-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR