3'-Cyano-2-phenylacetophenone ≥97%

Supplier: Matrix Scientific

086696-1G 086696-2G 086696-5G
102850-078EA 886.15 USD
102850-078 102850-080 102850-082
3'-Cyano-2-phenylacetophenone ≥97%
3'-Cyano-2-phenylacetophenone

Hazard: Irritant

Formula: C₁₅H₁₁NO
Melting Pt: 83…85 °C
MDL Number: MFCD02260674
CAS Number: 758710-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR