2'-Methoxy-2-phenylacetophenone ≥97%

Supplier: Matrix Scientific

086693-1G 086693-2G 086693-5G
102850-060EA 612.31 USD
102850-060 102850-062 102850-064
2'-Methoxy-2-phenylacetophenone ≥97%
2'-Methoxy-2-phenylacetophenone

Hazard: Irritant

Formula: C₁₅H₁₄O₂
MDL Number: MFCD02260671
CAS Number: 33470-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR