4'-Methyl-2-phenylacetophenone ≥97%

Supplier: Matrix Scientific

086692-1G 086692-2G 086692-5G
102850-054EA 192.31 USD
102850-054 102850-056 102850-058
4'-Methyl-2-phenylacetophenone ≥97%
4'-Methyl-2-phenylacetophenone

Hazard: Irritant

Formula: C₁₅H₁₄O
Melting Pt: 104…107 °C
MDL Number: MFCD00017215
CAS Number: 2001-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR