3'-Methyl-2-phenylacetophenone ≥97%
Supplier: Matrix Scientific
086691-1G
086691-2G
086691-5G
102850-048EA
347.69
USD
102850-048
102850-050
102850-052
3'-Methyl-2-phenylacetophenone ≥97%
3'-Methyl-2-phenylacetophenone
Hazard: Irritant
Formula:
C₁₅H₁₄O |
MDL Number:
MFCD02260670 CAS Number: 95606-81-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...