2'-Methyl-2-phenylacetophenone ≥97%
Supplier: Matrix Scientific
086690-1G
086690-2G
086690-5G
102850-042EA
550.77
USD
102850-042
102850-044
102850-046
2'-Methyl-2-phenylacetophenone ≥97%
2'-Methyl-2-phenylacetophenone
Hazard: Irritant
Formula:
C₁₅H₁₄O |
MDL Number:
MFCD02260669 CAS Number: 16216-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...