Benzyl-3-bromophenyl ketone ≥97%

Supplier: Matrix Scientific

086689-1G 086689-2G 086689-5G
102850-036EA 347.69 USD
102850-036 102850-038 102850-040
Benzyl-3-bromophenyl ketone ≥97%
Benzyl 3-bromophenyl ketone

Hazard: Irritant

Formula: C₁₄H₁₁BrO
MDL Number: MFCD02260666
CAS Number: 40396-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR