2'-Bromo-2-phenylacetophenone ≥97%
Supplier: Matrix Scientific
086688-1G
086688-2G
086688-5G
102850-030EA
520
USD
102850-030
102850-032
102850-034
2'-Bromo-2-phenylacetophenone ≥97%
2'-Bromo-2-phenylacetophenone
Hazard: Irritant
Formula:
C₁₄H₁₁BrO |
MDL Number:
MFCD01861486 CAS Number: 36081-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...