Benzyl-3-chlorophenyl ketone ≥97%

Supplier: Matrix Scientific

086686-1G 086686-2G 086686-5G
102850-018EA 347.69 USD
102850-018 102850-020 102850-022
Benzyl-3-chlorophenyl ketone ≥97%
Benzyl 3-chlorophenyl ketone

Hazard: Irritant

Formula: C₁₄H₁₁ClO
MDL Number: MFCD02260665
CAS Number: 62482-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR