2'-Chloro-2-phenylacetophenone ≥97%

Supplier: Matrix Scientific

086685-1G 086685-2G 086685-5G
102850-012EA 612.31 USD
102850-012 102850-014 102850-016
2'-Chloro-2-phenylacetophenone ≥97%
2'-Chloro-2-phenylacetophenone

Hazard: Irritant

Formula: C₁₄H₁₁ClO
MDL Number: MFCD02260664
CAS Number: 72867-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR