Benzyl-3-fluorophenyl ketone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(3-Fluorophenyl)-2-phenylethanone

086684-1G 086684-2G 086684-5G
102850-006EA 347.69 USD
102850-006 102850-008 102850-010
Benzyl-3-fluorophenyl ketone ≥97%
Benzyl 3-fluorophenyl ketone

Hazard: Irritant

Formula: C₁₄H₁₁FO
MW: 214.235 g/mol
MDL Number: MFCD02260663
CAS Number: 40281-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR