4-Chloro-1-oxo-1-(4-trifluoromethylphenyl)butane ≥97%

Supplier: Matrix Scientific

086682-1G 086682-2G 086682-5G
102849-994EA 592.31 USD
102849-994 102849-996 102849-998
4-Chloro-1-oxo-1-(4-trifluoromethylphenyl)butane ≥97%
4-Chloro-1-oxo-1-(4-trifluoromethylphenyl)butane

Hazard: Irritant

Formula: C₁₁H₁₀ClF₃O
MDL Number: MFCD02260641
CAS Number: 327617-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR