5-Chloro-1-(3-nitrophenyl)-1-oxopentane ≥97%

Supplier: Matrix Scientific

086681-1G 086681-2G 086681-5G
102849-988EA 347.69 USD
102849-988 102849-990 102849-992
5-Chloro-1-(3-nitrophenyl)-1-oxopentane ≥97%
5-Chloro-1-(3-nitrophenyl)-1-oxopentane

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₃
MDL Number: MFCD07699539
CAS Number: 487058-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR