5-Chloro-1-(4-nitrophenyl)-1-oxopentane ≥95%

Supplier: Matrix Scientific

086680-1G 086680-2G 086680-5G
102849-982EA 347.69 USD
102849-982 102849-984 102849-986
5-Chloro-1-(4-nitrophenyl)-1-oxopentane ≥95%
5-Chloro-1-(4-nitrophenyl)-1-oxopentane

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₃
MDL Number: MFCD08064679
CAS Number: 487058-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR