4-Chloro-1-(3-chlorophenyl)-1-oxobutane ≥97%
Supplier: Matrix Scientific
086679-1G
086679-2G
086679-5G
102849-976EA
347.69
USD
102849-976
102849-978
102849-980
4-Chloro-1-(3-chlorophenyl)-1-oxobutane ≥97%
4-Chloro-1-(3-chlorophenyl)-1-oxobutane
Hazard: Irritant
Formula:
C₁₀H₁₀Cl₂O |
MDL Number:
MFCD07699520 CAS Number: 90793-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...