4-Chloro-1-(3-fluorophenyl)-1-oxobutane ≥95%
Supplier: Matrix Scientific
086676-1G
086676-2G
086676-5G
102849-958EA
592.31
USD
102849-958
102849-960
102849-962
4-Chloro-1-(3-fluorophenyl)-1-oxobutane ≥95%
4-Chloro-1-(3-fluorophenyl)-1-oxobutane
Hazard: Irritant
Formula:
C₁₀H₁₀ClFO |
MDL Number:
MFCD02260552 CAS Number: 3110-52-9 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...