3-Chloro-1-(3-fluorophenyl)-1-oxopropane ≥97%

Supplier: Matrix Scientific

086675-1G 086675-2G 086675-5G
102849-952EA 592.31 USD
102849-952 102849-954 102849-956
3-Chloro-1-(3-fluorophenyl)-1-oxopropane ≥97%
3-Chloro-1-(3-fluorophenyl)-1-oxopropane

Hazard: Irritant

Formula: C₉H₈ClFO
MDL Number: MFCD07699514
CAS Number: 83406-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR