5-Chloro-1-(4-fluorophenyl)-1-oxopentane ≥97%
Supplier: Matrix Scientific
086674-1G
086674-2G
086674-5G
102849-946EA
520
USD
102849-946
102849-948
102849-950
5-Chloro-1-(4-fluorophenyl)-1-oxopentane ≥97%
5-Chloro-1-(4-fluorophenyl)-1-oxopentane
Hazard: Irritant
Formula:
C₁₁H₁₂ClFO |
MDL Number:
MFCD01319941 CAS Number: 17135-46-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...