1-(4-Bromophenyl)-5-chloro-1-oxopentane ≥97%
Supplier: Matrix Scientific
086673-1G
086673-2G
086673-5G
102849-940EA
592.31
USD
102849-940
102849-942
102849-944
1-(4-Bromophenyl)-5-chloro-1-oxopentane ≥97%
1-(4-Bromophenyl)-5-chloro-1-oxopentane
Hazard: Irritant
Formula:
C₁₁H₁₂BrClO |
MDL Number:
MFCD02260551 CAS Number: 54874-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...