6-Chloro-1-oxo-1-phenylhexane ≥97%
Supplier: Matrix Scientific
086672-1G
086672-2G
086672-5G
102849-934EA
347.69
USD
102849-934
102849-936
102849-938
6-Chloro-1-oxo-1-phenylhexane ≥97%
6-Chloro-1-oxo-1-phenylhexane
Hazard: Irritant
Formula:
C₁₂H₁₅ClO |
MDL Number:
MFCD00672047 CAS Number: 946-01-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...