5-Chloro-1-phenyl-1-pentanone ≥97%

Supplier: Matrix Scientific

086671-1G 086671-2G 086671-5G
102849-928EA 520 USD
102849-928 102849-930 102849-932
5-Chloro-1-phenyl-1-pentanone ≥97%
5-Chloro-1-phenyl-1-pentanone

Hazard: Irritant

Formula: (C₆H₅)C(O)CH₂CH₂CH₂CH₂Cl
MW: 196.68 g/mol
Melting Pt: 48-50°C
CAS Number: 942-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR