3-(4-Methoxybenzoyl)pyridine ≥97%
Supplier: Matrix Scientific
086648-1G
086648-2G
086648-5G
102849-796EA
795.38
USD
102849-796
102849-798
102849-800
3-(4-Methoxybenzoyl)pyridine ≥97%
3-(4-Methoxybenzoyl)pyridine
Hazard: Irritant
Formula:
C₁₃H₁₁NO₂ |
MDL Number:
MFCD01716433 CAS Number: 23826-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...