2-Methoxy-5-thiazoylpyridine ≥97%

Supplier: Matrix Scientific

086635-1G 086635-2G 086635-5G
102849-724EA 795.38 USD
102849-724 102849-726 102849-728
2-Methoxy-5-thiazoylpyridine ≥97%
2-Methoxy-5-thiazoylpyridine

Hazard: Irritant

Formula: C₁₀H₈N₂O₂S
MDL Number: MFCD07699468
CAS Number: 898785-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR