(2-Chloropyridin-4-yl)(cyclopropyl)methanone ≥97%
Supplier: Matrix Scientific
086634-1G
086634-2G
086634-5G
102849-718EA
795.38
USD
102849-718
102849-720
102849-722
(2-Chloropyridin-4-yl)(cyclopropyl)methanone ≥97%
(2-Chloropyridin-4-yl)(cyclopropyl)methanone
Hazard: Irritant
Formula:
C₉H₈ClNO |
MDL Number:
MFCD07699431 CAS Number: 898785-12-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...