2-(2,4-Dimethylbenzoyl)oxazole ≥97%

Supplier: Matrix Scientific

086632-1G 086632-2G 086632-5G
102849-706EA 795.38 USD
102849-706 102849-708 102849-710
2-(2,4-Dimethylbenzoyl)oxazole ≥97%
2-(2,4-Dimethylbenzoyl)oxazole

Hazard: Irritant

Formula: C₁₂H₁₁NO₂
MDL Number: MFCD07699395
CAS Number: 898784-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR