2-(4-Nitrobenzoyl)oxazole ≥97%

Supplier: Matrix Scientific

086629-1G 086629-2G 086629-5G
102849-688EA 795.38 USD
102849-688 102849-690 102849-692
2-(4-Nitrobenzoyl)oxazole ≥97%
2-(4-Nitrobenzoyl)oxazole

Hazard: Irritant

Formula: C₁₀H₆N₂O₄
Melting Pt: 1221 °C
MDL Number: MFCD07782996
CAS Number: 602295-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR