2-(3-Cyanobenzoyl)oxazole ≥97%
Supplier: Matrix Scientific
086627-1G
086627-2G
086627-5G
102849-676EA
795.38
USD
102849-676
102849-678
102849-680
2-(3-Cyanobenzoyl)oxazole ≥97%
2-(3-Cyanobenzoyl)oxazole
Hazard: Irritant
Formula:
C₁₁H₆N₂O₂ |
MDL Number:
MFCD07699357 CAS Number: 898759-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...