2-(3-Bromobenzoyl)oxazole ≥97%
Supplier: Matrix Scientific
086626-1G
086626-2G
086626-5G
102849-670EA
795.38
USD
102849-670
102849-672
102849-674
2-(3-Bromobenzoyl)oxazole ≥97%
2-(3-Bromobenzoyl)oxazole
Hazard: Irritant
Formula:
C₁₀H₆BrNO₂ |
MDL Number:
MFCD07699352 CAS Number: 898759-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...