2-(4-Chlorobenzoyl)oxazole ≥97%
Supplier: Matrix Scientific
086625-1G
086625-2G
086625-5G
102849-664EA
795.38
USD
102849-664
102849-666
102849-668
2-(4-Chlorobenzoyl)oxazole ≥97%
2-(4-Chlorobenzoyl)oxazole
Hazard: Irritant
Formula:
C₁₀H₆ClNO₂ |
MDL Number:
MFCD07699350 CAS Number: 898759-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...