2-(3-Chlorobenzoyl)oxazole ≥97%
Supplier: Matrix Scientific
086624-1G
086624-2G
086624-5G
102849-658EA
795.38
USD
102849-658
102849-660
102849-662
2-(3-Chlorobenzoyl)oxazole ≥97%
2-(3-Chlorobenzoyl)oxazole
Hazard: Irritant
Formula:
C₁₀H₆ClNO₂ |
MDL Number:
MFCD07699349 CAS Number: 898759-75-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...