2-(4-Fluorobenzoyl)oxazole ≥97%

Supplier: Matrix Scientific

086623-1G 086623-2G 086623-5G
102849-652EA 795.38 USD
102849-652 102849-654 102849-656
2-(4-Fluorobenzoyl)oxazole ≥97%
2-(4-Fluorobenzoyl)oxazole

Hazard: Irritant

Formula: C₁₀H₆FNO₂
MDL Number: MFCD07699347
CAS Number: 898759-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR