2-(4-Methoxybenzoyl)oxazole ≥97%

Supplier: Matrix Scientific

086619-1G 086619-2G 086619-5G
102849-628EA 1158.46 USD
102849-628 102849-630 102849-632
2-(4-Methoxybenzoyl)oxazole ≥97%
2-(4-Methoxybenzoyl)oxazole

Hazard: Irritant

Formula: C₁₁H₉NO₃
MDL Number: MFCD07699338
CAS Number: 898759-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR