2-(3-Methoxybenzoyl)oxazole ≥97%
Supplier: Matrix Scientific
086618-1G
086618-2G
086618-5G
102849-622EA
795.38
USD
102849-622
102849-624
102849-626
2-(3-Methoxybenzoyl)oxazole ≥97%
2-(3-Methoxybenzoyl)oxazole
Hazard: Irritant
Formula:
C₁₁H₉NO₃ |
MDL Number:
MFCD07699337 CAS Number: 898759-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...