2-(4-Chlorobutyryl)oxazole ≥95%
Supplier: Matrix Scientific
086615-1G
086615-2G
086615-5G
102849-604EA
795.38
USD
102849-604
102849-606
102849-608
2-(4-Chlorobutyryl)oxazole ≥95%
2-(4-Chlorobutyryl)oxazole
Hazard: Irritant
Formula:
C₇H₈ClNO₂ |
MDL Number:
MFCD07699306 CAS Number: 898758-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...