2-(4-tert-Butylbenzoyl)pyridine ≥97%
Supplier: Matrix Scientific
086614-1G
086614-2G
086614-5G
102849-598EA
795.38
USD
102849-598
102849-600
102849-602
2-(4-tert-Butylbenzoyl)pyridine ≥97%
2-(4-tert-Butylbenzoyl)pyridine
Hazard: Irritant
Formula:
C₁₆H₁₇NO |
MDL Number:
MFCD07699248 CAS Number: 157592-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...