2-(4-Nitrobenzoyl)pyridine ≥97%

Supplier: Matrix Scientific

086613-1G 086613-2G 086613-5G
102849-592EA 1158.46 USD
102849-592 102849-594 102849-596
2-(4-Nitrobenzoyl)pyridine ≥97%
2-(4-Nitrobenzoyl)pyridine

Hazard: Irritant

Formula: C₁₂H₈N₂O₃
Melting Pt: 99…100 °C
MDL Number: MFCD00957340
CAS Number: 27693-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR